missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Fc epsilon Receptor I alpha (FceR1) FITC, Clone: MAR-1, Invitrogen™
Descripción
Fc epsilon Receptor I alpha (FceR1) Monoclonal antibody A18400 from Invitrogen specifically detects Fc epsilon Receptor I alpha (FceR1) in Mouse samples. It is validated for Flow Cytometry.
The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
Host Species: Hamster
Cross Reactivity: Mouse
Isotype: IgG
Clone: MAR-1
Immunogen: mouse Fc epsilon receptor I (FceR1) alpha subunit
Formulation: 50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: FCRG, Fc receptor gamma-chain, fc-epsilon RI-gamma, fcRgamma, fceRI gamma, high affinity immunoglobulin epsilon receptor subunit gamma, immunoglobulin E receptor, high affinity, gamma chain
Uniprot Accession: P20489
Entrez GeneID: 14125
Gene Symbol: Fcer1a
This product detects Fc epsilon Receptor I alpha (FceR1) in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is mouse Fc epsilon receptor I (FceR1) alpha subunit.
Host Species: Hamster
Cross Reactivity: Mouse
Isotype: IgG
Clone: MAR-1
Immunogen: mouse Fc epsilon receptor I (FceR1) alpha subunit
Formulation: 50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: FCRG, Fc receptor gamma-chain, fc-epsilon RI-gamma, fcRgamma, fceRI gamma, high affinity immunoglobulin epsilon receptor subunit gamma, immunoglobulin E receptor, high affinity, gamma chain
Uniprot Accession: P20489
Entrez GeneID: 14125
Gene Symbol: Fcer1a
This product detects Fc epsilon Receptor I alpha (FceR1) in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is mouse Fc epsilon receptor I (FceR1) alpha subunit.
Especificaciones
Especificaciones
| Antígeno | Fc epsilon Receptor I alpha (FceR1) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | MAR-1 |
| Conjugado | FITC |
| Formulación | 50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide |
| N.º de referencia del gen | P20489 |
| Alias de gen | FCRG, Fc receptor gamma-chain, fc-epsilon RI-gamma, fcRgamma, fceRI gamma, high affinity immunoglobulin epsilon receptor subunit gamma, immunoglobulin E receptor, high affinity, gamma chain |
| Símbolos de los genes | Fcer1a |
| Especie del huésped | Hamster |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?