missing translation for 'onlineSavingsMsg'
Learn More

anti-FAS/CD95 PE, Clone: 15A7, Invitrogen™

Código de producto. 15206074
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15206074 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15206074 Proveedor Invitrogen N.º de proveedor A16388

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

FAS/CD95 Monoclonal antibody A16388 from Invitrogen specifically detects FAS/CD95 in Mouse samples. It is validated for Flow Cytometry.

Fas ligand (FasL, CD 95L) is a type-II-membrane protein, whose N-terminus is in the cytoplasm and its C-terminal region extends into the extracellular space. Its receptor Fas (CD 95, Apo-1) is a cell-surface-type-I-membrane protein and a member of the tumor necrosis factor (TNF) and nerve growth factor (NGF) receptor family. As a member of the TNF-cytokine family FasL induces apoptosis when interacting with its receptor Fas. FasL may exist as either membrane bound (45 kD) or soluble forms (26 kD). The soluble protein can be released from cells upon cleavage by metalloproteinases. Binding of FasL to Fas leads to oligomerization of the receptor and triggers apoptotic cell death through the interaction of other proteins. FasL is predominantly expressed in activated T-lymphocytes and natural killer (NK) cells also it is expressed in the tissues of immune-privilege sites such as testis and eye. FasL expression is also reported in various tissues as thymus, liver, ovary, lung, heart and kidney. The Fas/FasL system has been shown to play a role in a number of human diseases, for example AIDS, hepatitis or cancer. It is assumed that induction of apoptosis through FasL is predominantly involved in anti-viral immune responses.
Host Species: Mouse
Cross Reactivity: Mouse
Isotype: IgG1, κ
Clone: 15A7
Immunogen: Mouse CD95 Protein
Formulation: 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Uniprot Accession: P25446
Entrez GeneID: 14102
Gene Symbol: Fas
This product detects FAS/CD95 in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse CD95 Protein.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FAS/CD95
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 15A7
Conjugado PE
Formulación 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
N.º de referencia del gen P25446
Alias de gen ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Símbolos de los genes Fas
Especie del huésped Mouse
Inmunógeno Mouse CD95 Protein
Cantidad 50 μg
Primario o secundario Primary
ID de gen (Entrez) 14102
Especies diana Mouse
Contenido y almacenamiento Store at 4° C
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.