missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR1 Polyclonal Antibody, Invitrogen™
Descripción
Recommended positive controls: Jurkat, Raji, K562, C8D30, NIH-3T3, Raw264.7, C2C12, rat liver. Predicted reactivity: Mouse (86%), Rat (84%), Dog (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EMR1 has a domain resembling seven transmembrane G protein-coupled hormone receptors (7TM receptors) at its C-terminus. The N-terminus of the encoded protein has six EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. EMR1 is the human ortholog of murine F4/80 but unlike F4/80, the expression of EMR1 in humans is restricted to eosinophils and is not expressed on macrophage populations.
Especificaciones
Especificaciones
| Antígeno | EMR1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.23 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| génica | ADGRE1 |
| N.º de referencia del gen | Q14246, Q5Y4N8, Q61549 |
| Alias de gen | ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; egf-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 |
| Símbolos de los genes | ADGRE1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?