missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CXCR4, PE, Clone: 44717, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB173P-025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry.Especificaciones
| CXCR4 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CXCR4 | |
| Mouse 3T3 cells transfected with human CXCR4 | |
| 25 Tests | |
| 7852 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 44717 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Reacts specifically with human and non-human cells expressing human CXCR4 (fusin) as detected by flow cytometry. It will also react with cells expressing feline CXCR4 but not rat CXCR4. This antibody does not cross-react with other chemokine receptors. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?