Learn More
CEACAM1, Human, Matched Antibody Pair (AP42), Abnova™
Código de producto. 16121784
Change view
Click to view available options
Cantidad:
1 Set
Tamaño de la unidad:
1 set
16121784 1 Set 1 set
1 options
Código de producto. 16121784 Proveedor Abnova N.º de proveedor H00000634AP42.1Set
Includes
Capture antibody: mouse monoclonal anti-CEACAM1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CEACAM1, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Especificaciones
Antígeno CEACAM1
Aplicaciones ELISA
Clasificación Monoclonal/Polyclonal
Conjugado Biotin
Descripción This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CEACAM1.
génica CEACAM1
Alias de gen BGP/BGP1/BGPI
Símbolos de los genes CEACAM1
Especie del huésped Mouse
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 634
Incluye Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CEACAM1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CEACAM1, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only