Learn More
anti-CDX4, Human, Matched Antibody Pair, Abnova™
Código de producto. 16121884
Change view
Click to view available options
Cantidad:
1 Set
Tamaño de la unidad:
1 set
16121884 1 Set 1 set
1 options
Código de producto. 16121884 Proveedor Abnova N.º de proveedor H00001046AP41.1Set
Includes
Capture antibody: mouse monoclonal anti-CDX4, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CDX4, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Especificaciones
Antígeno CDX4
Aplicaciones ELISA
Clasificación Monoclonal/Polyclonal
Conjugado Biotin
Descripción This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CDX4.
génica CDX4
Símbolos de los genes CDX4
Especie del huésped Mouse
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1046
Incluye Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CDX4, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CDX4, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only