Learn More
CD69 Hamster anti-Mouse, PE-CF594, Clone: H1.2F3, BD
Descripción
The H1.2F3 monoclonal antibody specifically binds to CD69 (Very Early Activation antigen), an 85 kDa disulfide-linked homodimer of differentially glycosylated subunits. CD69 is a C-type lectin, most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. Its expression is rapidly induced upon activation of lymphocytes (T, B, NK, and NK-T cells), neutrophils, and macrophages. CD69 is expressed also on thymocytes that are undergoing positive selection; its role in that process is unclear. H1.2F3 mAb augments PMA-induced T-cell stimulation and IFN- γ-induced macrophage stimulation. IL-2-activated NK cells express CD69, and H1.2F3 mAb induces redirected lysis of FcR-bearing target cells by NK cells.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Especificaciones
Especificaciones
| Antígeno | CD69 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | H1.2F3 |
| Concentración | 0.2mg/mL |
| Conjugado | PE-CF594 |
| Descripción | VEA; Very Early Activation Antigen; AIM; Activation Induced Molecule |
| Formulación | Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. |
| Especie del huésped | Hamster |
| Inmunógeno | Mouse Dendritic Epidermal T Cell Line Y245 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.