Learn More
CD68 Mouse anti-Human, PE-CF594, Clone: Y1/82A, BD
Descripción
The Y1/82A monoclonal antibody specifically binds to CD68 which is also known as, Scavenger receptor class D, member 1 (SCARD1), Macrosialin, or GP110. CD68 is a cell surface 110 kDa-type I-transmembrane glycoprotein that is primarily expressed in cytoplasmic granules of monocytes, macrophages, dendritic cells, granulocytes, myeloid progenitor cells and, reportedly, a subset of CD34-positive hemopoietic bone marrow progenitor cells. CD68 belongs to the sialomucin family and serves as a scavenger receptor that can bind and internalize oxidized low density lipoproteins (LDL). This antibody is useful in studies of myeloid cell development and function.
This antibody is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Especificaciones
Especificaciones
| Antígeno | CD68 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | Y1/82A |
| Conjugado | PE-CF594 |
| Descripción | GP110; Macrosialin; SCARD1; Scavenger receptor class D, member 1 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
| Especie del huésped | Mouse |
| Inmunógeno | PHA-stimulated Human PBMC |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.