Learn More
CD154 (CD40 Ligand), FITC, clone: 40804, Invitrogen™
Código de producto. 15553272 Tienda Thermo Scientific Productos
Cambiar vista
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
100 tests
15553272 100 Tests 100 tests
1 options
Código de producto. 15553272 Proveedor Invitrogen N.º de proveedor MA523582
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Mouse Monoclonal Antibody
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD27 Ligand, rhCD30 Ligand, or recombinant mouse CD40 Ligand is observed.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD154 (CD40 Ligand)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 40804
Conjugado FITC
Formulación Buffered saline solution with BSA and 0.09% sodium azide
génica CD40LG
N.º de referencia del gen P29965
Alias de gen RP23-153G22.3, CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39, T-cell antigen Gp39, TNF-related activation protein, tumor necrosis factor ligand superfamily member 5
Símbolos de los genes CD40LG
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human CD40 Ligand/TNFSF5 Leu50-Leu261
Método de purificación Protein A/G
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 959
Especies diana Human
Contenido y almacenamiento Store at 4°C, store in the dark, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b