Learn More
CD3E, Human, Matched Antibody Pair (AP41), Abnova™
Código de producto. 16053738
Change view
Click to view available options
Cantidad:
1 Set
Tamaño de la unidad:
1 set
16053738 1 Set 1 set
1 options
Código de producto. 16053738 Proveedor Abnova N.º de proveedor H00000916AP41.1Set
Includes
Capture antibody: mouse monoclonal anti-CD3E (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Especificaciones
Antígeno CD3E
Aplicaciones ELISA
Clasificación Monoclonal/Polyclonal
Conjugado Biotin
Descripción This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD3E.
génica CD3E
Alias de gen FLJ18683/T3E/TCRE
Símbolos de los genes CD3E
Especie del huésped Mouse
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 916
Incluye Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD3E (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only