missing translation for 'onlineSavingsMsg'
Learn More

anti-CD3e/CD3 epsilon FITC, Clone: 145-2C11, Invitrogen™

Código de producto. 15434833
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15434833 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15434833 Proveedor Invitrogen N.º de proveedor A27060

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

CD3e/CD3 epsilon Monoclonal antibody A27060 from Invitrogen specifically detects CD3e/CD3 epsilon in Mouse samples. It is validated for Flow Cytometry.

CD3 complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases.
Host Species: Armenian Hamster
Cross Reactivity: Mouse
Isotype: IgG
Clone: 145-2C11
Immunogen: Mouse CD3 epsilon
Formulation: 100μg Purified Antibody in PBS, with 0.1% gelatin and 0.09% sodium azide
Synonyms: FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Uniprot Accession: P22646
Entrez GeneID: 12501
Gene Symbol: Cd3e
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD3e/CD3 epsilon
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 145-2C11
Conjugado FITC
Formulación 100μg Purified Antibody in PBS, with 0.1% gelatin and 0.09% sodium azide
N.º de referencia del gen P22646
Alias de gen FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Símbolos de los genes Cd3e
Especie del huésped Armenian Hamster
Inmunógeno Mouse CD3 epsilon
Cantidad 100 μg
Primario o secundario Primary
ID de gen (Entrez) 12501
Especies diana Mouse
Contenido y almacenamiento Store at 4°C, in the dark
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.