Learn More
CD11b/Mac-1 Mouse anti-Human, Alexa Fluor 700, Clone: ICRF44, BD
Descripción
The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.
This antibody conjugate is routinely tested by flow cytometry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Especificaciones
Especificaciones
| Antígeno | CD11b |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | ICRF44 |
| Concentración | 0.2mg/mL |
| Conjugado | Alexa Fluor 700 |
| Descripción | Mac-1α, integrin αM subunit, CR3 α chain |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human monocytes |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.