missing translation for 'onlineSavingsMsg'
Learn More

CD11a Mouse anti-Human, FITC, Clone: HI111, BD

Código de producto. 15868118
Change view
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15868118 100 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15868118 Proveedor BD Biosciences N.º de proveedor 555383

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD11a
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon HI111
Conjugado FITC
Descripción LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 100 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Canine, Human, Rabbit
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.