missing translation for 'onlineSavingsMsg'
Learn More

anti-C1QB Matched Antibody Pair, Abnova™

Código de producto. 16023468
Change view
Click to view available options
Cantidad:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16023468 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16023468 Supplier Abnova Supplier No. H00000713AP21.1Set

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Includes

Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-C1QB (100μg); Detection antibody: mouse purified polyclonal anti-C1QB, (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human C1QB

Host Species: Rabbit, Mouse
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Specifications

Antígeno C1QB
Aplicaciones ELISA
Clasificación Monoclonal/Polyclonal
Conjugado Unconjugated
Descripción This antibody pair set comes with matched antibody pair to detect and quantify protein level of human C1QB.
génica C1QB
Símbolos de los genes C1QB
Especie del huésped Mouse, Rabbit
Método de purificación Affinity Purified
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 713
Incluye Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-C1QB (100 μg) 2. Detection antibody: mouse purified polyclonal anti-C1QB (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.