Learn More
BAF60a Mouse, Unlabeled, Clone: 23, BD
Descripción
SWI/SNF complexes facilitate gene activation and transcription factor binding byaltering repressive chromatin structures. In mammals, SWI/SNF complexes are present inmutliple forms that include 9-12 proteins called B RG1- a ssociated f actors (BAFs), whichrange in molecular weight from 47 to 250 kDa. BAF60 is the homologue of the yeastSWP70 subunit of the yeast SWI/SNF complex that regulates glucocorticoidtranscription. Three isoforms of BAF60 are BAF60a, BAF60b, and BAF60c. BAF60a isubiquitously expressed and is present in the 200-kDa BRG1 complex. BAF60b andBAF60c are expressed preferentially in muscle and pancreas and may be subunits of adistinct complex that shares some subunits with BRG1 complex. In hematopoietic cells,a SWI/SNF related-complex (PYR) contains BAF57, INI1, BAF60a, and BAF170. This complex binds a 250 bp pyrimidine-rich element between the fetal and adult ß-globingenes and may facilitate human fetal-to-adult globin gene switching. Thus, BAF60proteins are components of multiple SWI/SNF complexes that regulate the transcriptionof various genes.
Especificaciones
Especificaciones
| Antígeno | BAF60a |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 23 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse BAF60a, aa 1-91 |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.