missing translation for 'onlineSavingsMsg'
Learn More

alpha-1 Antitrypsin Mouse anti-Human, Clone: 3B7, Invitrogen™

Código de producto. 11564342
Cambiar vista
Click to view available options
Cantidad:
200 μL
Tamaño de la unidad:
200 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11564342 200 μL 200 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11564342 Proveedor Invitrogen N.º de proveedor HYB1910102

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

HYB 191-01-02 has specificity for human a-1 AT, native and denatured.   HYB 191-01-02 has been successfully used in Western blotting and ELISA procedures. HYB 191-01-02 reacts strongly with a-1 AT. Strong reaction is seen in ELISA with native and denatured a-1 AT directly coated onto the microtiter well. When tested in sandwich ELISA in combination with a polyclonal antibody against a-1 AT (e.g. DAKO A0012), it only recognizes native a-1 AT. In Western blotting after SDS-PAGE, HYB 191-01-02 reacts with native and denatured a-1 AT in reduced as well as non-reduced forms.  The HYB 191-01-02 immunogen is a-1 AT isolated from human plasma. The HYB 191-01-02 epitope differs from that of HYB 185-02-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL

The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno alpha-1 Antitrypsin
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon 3B7
Concentración 1.05 mg/mL
Conjugado Unconjugated
génica SERPINA1
N.º de referencia del gen P01009
Alias de gen a-1 AT
Símbolos de los genes SERPINA1
Especie del huésped Mouse
Inmunógeno a-1 AT isolated from human plasma.
Método de purificación Protein A
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5265
Especies diana Human
Contenido y almacenamiento 4° C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.