missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ankyrin 1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15701152 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15701152 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15701152 Proveedor Invitrogen™ N.º de proveedor PA563372

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: FVLVSDKHRM SFPETVDEIL DVSEDEGEEL ISFKAERRDS RDVDEEKELL DFVPKLDQVV ESPAIPRIPC AMPETVVIRS EEQEQASKEY DEDSLIPSSP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 83%.

Ankyrins are a family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton and play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Multiple isoforms of ankyrin with different affinities for various target proteins are expressed in a tissue-specific, developmentally regulated manner. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. Ankyrin 1, the prototype of this family, was first discovered in the erythrocytes, but since has also been found in brain and muscles. Mutations in erythrocytic ankyrin 1 have been associated in approximately half of all patients with hereditary spherocytosis. Complex patterns of alternative splicing in the regulatory domain, giving rise to different isoforms of ankyrin 1 have been described. Truncated muscle-specific isoforms of ankyrin 1 resulting from usage of an alternate promoter have also been identified.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Ankyrin 1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.6 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica ANK1
N.º de referencia del gen P16157
Alias de gen ANK; ANK1; ANK-1; ANK-1; erythrocyte skeletal protein; ankyrin 1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin-1; Ankyrin-R; erythrocyte ankyrin; nb; normoblastic anemia; pale; rCG_43073; SPH1; SPH2
Símbolos de los genes ANK1
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human Ankyrin 1. Recombinant protein control fragment (Product #RP-100186).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 286
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.