Learn More
Tocris Bioscience™ Ani 9
Potent and selective TMEM16A (Anoctamin-1; ANO1) blocker
Marca: Tocris Bioscience™ 6076/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ani 9 is a potent and selective TMEM16A (Anoctamin-1; ANO1) blocker (IC50 = 77 nM). Displays no effect on ANO2 activity (at 10 μM). Also exhibits no effect on intracellular calcium signaling or CFTR and ENaC channel activity (at 30 μM).Especificaciones
| Ani 9 | |
| 2-(4-Chloro-2-methylphenoxy)-N'-(2-methoxybenzylidene)acetohydrazide | |
| Soluble to 100mM in DMSO | |
| Off-white |
| 356102-14-2 | |
| 50 mg | |
| Solid | |
| Bioactive Small Molecule |
Título del producto
For Research Use Only.