Learn More
MedChemExpress Angiotensin (1-7)
Descripción
Angiotensin 1-7 inhibits purified canine ACE activity (IC50=0.65 μM). Angiotensin 1-7 acts as a local synergistic modulator of kinin-induced vasodilation by inhibiting ACE and releasing nitric oxide. Angiotensin 1-7 blocks Ang II-induced smooth muscle cell proliferation and hypertrophy and shows antiangiogenic and growth-inhibitory effects on the endothelium. Angiotensin 1-7 shows anti-inflammatory activity .
Especificaciones
Especificaciones
| Pureza | 98.04% |
| CAS | 51833-78-4 |
| Contenido y almacenamiento | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Para utilizar con (aplicación) | COVID-19-anti-virus |
| Peso molecular | 899.0 g/mol |
| Tipo de producto | Peptides |
| Cantidad | 5 mg |
| Condiciones de envío | Room temperature |
| Solubilidad | H2O : ≥ 30.2 mg/mL (33.59 mM) |
| Sinónimo | Ang-(1-7) |