Learn More
Tocris Bioscience™ Angiopep-2
Descripción
Angiopep-2 is a synthetic peptide able to specifically interact with low-density lipoprotein receptor-related protein 1 (LRP1) receptors present both on blood-brain barrier (BBB) and tumor cell membranes. Angiopep-2 binding to LRP1 facilitates receptor-mediated transcytosis of cargo across the BBB. Transcytosis-inducing molecular degraders of extracellular proteins (TransMoDEs), containing Angiopep-2 and modified to include a small-molecule ligand, induce target internalization that can cross the BBB in a variety of cell types.
Especificaciones
Especificaciones
| CAS | 906480-05-5 |
| Cantidad | 1 mg |
| Objetivo | LDL Receptor Peptides |
| Fórmula molecular | C104H149N29O31 |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | ≥95% (HPLC) |
| Peso molecular | 2301.5 |