Learn More
Anagliptin, MedChemExpress
Código de producto. 16498817
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16498817 5 mg 5 miligramos
16466228 10 mg 10 miligramos
16442357 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16498817 Proveedor Medchem Express N.º de proveedor HY14877
MedChemExpress Anagliptin (SK-0403) is a highly selective, potent, orally active inhibitor of dipeptidyl peptidase 4 (DPP-4), with an IC50 of 3.8 nM, and less selective at DPP-8 and DDP-9 with IC50s of 68 nM and 60 nM, respectively.
Anagliptin (SK-0403) is a highly selective, potent, orally active inhibitor of dipeptidyl peptidase 4 (DPP-4), with an IC50 of 3.8 nM, and less selective at DPP-8 and DDP-9 with IC50s of 68 nM and 60 nM, respectively.
Especificaciones
Nombre del producto químico o material Anagliptin
CAS 739366-20-2
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C19H25N7O2
Cantidad 5 mg
Sinónimo SK-0403
Información de solubilidad DMSO : 100 mg/mL (260.79 mM; Need ultrasonic)
SMILES O=C(C1=CN2C(N=C1)=CC(C)=N2)NCC(C)(NCC(N3[C@H](C#N)CCC3)=O)C
Peso molecular (g/mol) 383.45
Formula Weight (peso de la fórmula) 383.45
Porcentaje de pureza 97.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism