Learn More
MedChemExpress β-Amyloid (1-40) (rat)
Código de producto. 16462889
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
16462889 1 mg 1 miligramo
1 options
Código de producto. 16462889 Proveedor MedChemExpress N.º de proveedor HYP1387
MedChemExpress β-Amyloid (1-40) (rat) is a rat form of the amyloid β-peptide, which accumulates as an insoluble extracellular deposit around neurons, giving rise to the senile plaques associated with Alzheimer's disease (AD).
β-Amyloid (1-40) (rat) increases 45Ca2+ influx, induces neurodegeneration in the rat hippocampal neurons of the CA1 subfield. β-Amyloid (1-40) (rat) induces apoptosis. β-Amyloid (1-40) (rat) can be used for the research of Alzheimer's disease.
Especificaciones
CAS 144409-98-3
Contenido y almacenamiento Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Para utilizar con (aplicación) Neuroscience-Neurodegeneration
Peso molecular 4233.76 g/mol
Tipo de producto Peptides