Learn More
Succinato de amonio, Thermo Scientific Chemicals
Código de producto. 17241282 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Cantidad:
5 g
25 g
Tamaño de la unidad:
25 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17241282 5 g 5 g
17251282 25 g 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17241282 Proveedor Thermo Scientific Acros N.º de proveedor 466860050
Solicitar formato a granel o personalizado
CAS: 2226-88-2 | C4H12N2O4 | 152.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2226-88-2
Fórmula molecular C4H12N2O4
Peso molecular (g/mol) 152.15
Clave InChI NHJPVZLSLOHJDM-UHFFFAOYSA-N
Nombre IUPAC Butanedioato de diammonio
SMILES [NH4+].[NH4+].[O-]C(=O)CCC([O-])=O
Tekniske data
Nombre del producto químico o material Ammonium succinate
Cantidad 5 g
Formula Weight (peso de la fórmula) 152.12
RUO – Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.