Learn More
Tocris Bioscience™ AMG PERK 44
Descripción
AMG PERK 44 is a potent and selective PERK inhibitor (IC50 = 6 nM). Exhibits >160-fold selectivity for PERK over B-Raf and GCN2, as well as a panel of 387 other kinases. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride |
| CAS | 1883548-84-2 |
| Cantidad | 10 mg |
| Objetivo | PERK Inhibitors |
| Fórmula molecular | C34H28N4O2.HCl |
| Formula Weight (peso de la fórmula) | 561.07 |
| Pureza | >98% |