Learn More
Ambroxol, MedChemExpress
Código de producto. 16448576
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100 mg
Tamaño de la unidad:
100 miligramos
16448576 100 mg 100 miligramos
1 options
Código de producto. 16448576 Proveedor Medchem Express N.º de proveedor HYB1039
MedChemExpress Ambroxol (NA-872), an active metabolite of the prodrug Bromhexine, has potent expectorant effects. Ambroxol is a glucocerebrosidase (GCase) chaperone and increases glucocerebrosidase activity. Ambroxol induces lung autophagy and has the potential for Parkinson disease and neuronopathic Gaucher disease research.
Ambroxol (NA-872), an active metabolite of the prodrug Bromhexine, has potent expectorant effects. Ambroxol is a glucocerebrosidase (GCase) chaperone and increases glucocerebrosidase activity. Ambroxol induces lung autophagy and has the potential for Parkinson disease and neuronopathic Gaucher disease research.
Especificaciones
Nombre del producto químico o material Ambroxol
CAS 18683-91-5
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C13H18Br2N2O
Cantidad 100 mg
Sinónimo NA-872
Información de solubilidad DMSO : ≥ 200 mg/mL (528.96 mM)
SMILES O[C@H]1CC[C@H](NCC2=CC(Br)=CC(Br)=C2N)CC1
Peso molecular (g/mol) 378.1
Formula Weight (peso de la fórmula) 378.1
Porcentaje de pureza 99.83%
Grado Research
Forma física Solid