Learn More
Tocris Bioscience™ AM 4113
Selective, high affinity CB1 antagonist
Marca: Tocris Bioscience™ 6193/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
AM 4113 is a high affinity and selective CB1 antagonist (Kd = 0.89 nM). Exhibits 100-fold selectivity for CB1 over CB2. Attenuates CB1 agonist AM 411-induced locomotor suppression and reduces food intake in vivo, but does not induce signs of nausea.Especificaciones
| 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide | |
| 614726-85-1 | |
| >98% | |
| 10 mg |