Learn More
Glicinato de aluminio, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | Aluminum Glycinate |
| CAS | 13682-92-3 |
| Fórmula molecular | C2H6AlNO4 |
| Cantidad | 25 g |
| Sinónimo | (Glycinato)dihydroxoaluminum, (Glycinato-N,O)dihydroxyaluminum, Ada, Alamine, Aldimate, Alglyn, Alminate, Alubasine, Aluminum Dihydroxyaminoacetate, Alzinox, Aspogen, Basic Aluminum Glycinate, Dihydroxyaluminum Aminoacetate, Dihydroxyaluminum Glycinate, Dimothyn, Doraxamin, Elcosal, Glycinal, Robalate |
| SMILES | O=C1C[NH2][Al](O)(O)O1 |
| Peso molecular (g/mol) | 135.05 |