Learn More
Altretamine, MedChemExpress
Product Code. 16458156
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 mg
500 mg
Unit Size:
100 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16458156 100 mg 100 miligramos
16422056 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458156 Supplier Medchem Express Supplier No. HYB0181
This item is not returnable. View return policy
MedChemExpress Altretamine is an alkylating antineoplastic agent.
Altretamine is an alkylating antineoplastic agent.
Specifications
Nombre del producto químico o material Altretamine
Forma física Solid
CAS 645-05-6
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C9H18N6
Para utilizar con (aplicación) Cancer-programmed cell death
Sinónimo ENT-50852 RB-1515 WR-95704
Información de solubilidad DMSO : 8.33 mg/mL (39.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CN(C)C1=NC(N(C)C)=NC(N(C)C)=N1
Peso molecular (g/mol) 210.28
Formula Weight (peso de la fórmula) 210.28
Porcentaje de pureza 99.64%