Learn More
alfa-terpineol-d3 (propilmetil-d3), CDN
Product Code. 30317997
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
0.05 g
0.1 g
missing translation for 'unitSize'
0.05 g
0.10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30317997 0.1 g 0.10 g
30314927 0.05 g 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317997 missing translation for 'mfr' CDN missing translation for 'supplierNo' CDND27070.1G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material alpha-Terpineol alpha-methyl-d3
CAS 203633-12-9
Fórmula molecular C10 2H3 H15 O
Cantidad 0.1 g
Sinónimo 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(methyl-d3)- (9 CI), α-Terpineol-d3, α-Terpineol-D3 (α-methyl D3)
Fórmula InChI InChI=1 S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11 H,5-7H2,1-3H3/i2D3
SMILES [2 H]C([2 H])([2 H])C(C)(O)C1CCC(=CC1)C
Nombre IUPAC 1,1,1-trideuterio-2-(4-metilciclohex-3-en-1-il)propan-2-ol
Peso molecular (g/mol) 157.27
Formula Weight (peso de la fórmula) 157.1546 g/mol
Porcentaje de pureza 98 atom % D, min 98% Chemical Purity