Learn More
Alpaca anti-Rabbit IgG Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Description
Biclonal nanobodies specific for rabbit IgG Fab and Fc. CTK0101: KD = 0.2 nM, CTK0102: KD = 1.2 nM Non Cross-Reactivity includes: Mouse, rat, sheep, goat, and guinea pig serum. Cross-reactivity to: Human and macaque serum Rapid workflow: Simultaneous incubation of rabbit IgG primary antibody and Alpaca anti-rabbit IgG Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek.
Specifications
Specifications
| Antígeno | Rabbit IgG Nano (VHH) |
| Aplicaciones | Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot |
| Clasificación | Monoclonal |
| Clon | CTK0101/CTK0102 |
| Concentración | 0.5 mg/mL |
| Conjugado | Alexa Fluor 647 |
| Formulación | 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7 |
| Alias de gen | Biclonal; Nanobodies; recombinant VHH |
| Especie del huésped | Alpaca |
| Inmunógeno | Purified rabbit IgG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.