Learn More
Alpaca anti-Mouse IgG2b Nano (VHH) Recombinant Secondary Antibody, Alexa Fluor™ 568, Invitrogen™
Código de producto. 17192234
Change view
Click to view available options
Cantidad:
10 μL
Tamaño de la unidad:
10 microlitros
17192234 10 μL 10 microlitros
1 options
Código de producto. 17192234 Proveedor Invitrogen N.º de proveedor SA510336
Alpaca Recombinant Monoclonal Antibody
Biclonal nanobodies specific for mouse IgG2b Fc. CTK0105: KD = 5 nM, CTK0106: KD = 0.2 nM Non Cross-Reactivity includes: Rabbit, rat, goat, sheep, guinea pig, human, and macaque serum and to mouse IgG1, IgG2a, IgG2c, and IgG3. Rapid workflow: Simultaneous incubation of mouse IgG2b primary antibody and Alpaca anti-mouse IgG2b Nano (VHH) recombinant secondary antibody. Offered through a partnership with ChromoTek
Mouse IgG2b Nano (VHH) Recombinant Monoclonal antibody specifically detects Mouse IgG2b Nano (VHH) in Mouse samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Mouse IgG2b Nano (VHH)
Aplicaciones Dot Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Clasificación Monoclonal
Clon CTK0105/CTK0106
Concentración 0.5 mg/mL
Conjugado Alexa Fluor 568
Formulación 10mM HEPES with 500mM NaCl, 5mM EDTA and 0.09% sodium azide; pH 7
Alias de gen Biclonal; Fc-specific recombinant VHH; Nanobodies
Especie del huésped Alpaca
Inmunógeno Purified mouse IgG2b
Método de purificación Affinity chromatography
Cantidad 10 μL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2b