Learn More
Alopurinol-d2, TRC
Product Code. 30322440
Change view
Click to view available options
Cantidad:
2.5 mg
Unit Size:
2.50 miligramo
Product Code. Cantidad unitSize
30322440 2.5 mg 2.50 miligramo
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322440 Supplier Toronto Research Chemicals Supplier No. TRCA5473022.5MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Allopurinol-d2
CAS 916979-34-5
Fórmula molecular C5 2H2 H2 N4 O
Cantidad 2.5 mg
Sinónimo 4H-Pyrazolo[3,4-d]pyrimidin-4-one-3,6-d2, 1,5-dihydro- (9CI), 1,5-Dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one-3,6-d2, 4-Hydroxypyrazolo[3,4-d]pyrimidine-3,6-d2, Allopurinol-3,6-D2, Allopurinol-d2
Fórmula InChI InChI=1S/C5H4N4O/c10-5-3-1-8-9-4(3)6-2-7-5/h1-2H,(H2,6,7,8,9,10)/i1D,2D
SMILES [2H]C1=NC(=O)c2c([2H])n[nH]c2N1
Nombre IUPAC 3,6-dideuterio-1,7-dihidropirazolo[3,4-d]pirimidin-4-ona
Peso molecular (g/mol) 138.12
Formula Weight (peso de la fórmula) 138.0511