Learn More
Aldicarb-d3, TRC
Product Code. 30309172
Change view
Click to view available options
Cantidad:
10 mg
Unit Size:
10 miligramos
Product Code. Cantidad unitSize
30309172 10 mg 10 miligramos
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309172 Supplier Toronto Research Chemicals Supplier No. TRCA51465210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Moléculas orgánicas de alta pureza y estándares analíticos, entregados estratégicamente en todo el mundo para potenciar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Aldicarb D3 (methyl-D3)
CAS 1795142-83-4
Fórmula molecular C7 H14 N2 O2 S
Cantidad 10 mg
Sinónimo [(2-Methyl-2-methylsulfanylpropylidene)amino] N-(trideuteriomethyl)carbamate
Fórmula InChI InChI=1S/C7H14N2O2S/c1-7(2,12-4)5-9-11-6(10)8-3/h5H,1-4H3,(H,8,10)/i3D3
SMILES [2H]C([2H])([2H])NC(=O)ON=CC(C)(C)SC
Nombre IUPAC [(2-metil-2-metilsulfanilpropilideno)amino] N-(trideuteriometil)carbamato
Peso molecular (g/mol) 193.28
Formula Weight (peso de la fórmula) 193.0964