Learn More
AKT inhibitor VIII, MedChemExpress
Código de producto. 16476587
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16476587 5 mg 5 miligramos
16434896 100 mg 100 miligramos
16430237 50 mg 50 miligramos
16403868 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16476587 Proveedor Medchem Express N.º de proveedor HY10355
MedChemExpress AKT inhibitor VIII (AKTi-1/2) is a cell-permeable quinoxaline compound that has been shown to potently, selectively, allosterically, and reversibly inhibit Akt1, Akt2, and Akt3 activity with IC50s of 58 nM, 210 nM, and 2119 nM, respectively.
AKT inhibitor VIII (AKTi-1/2) is a cell-permeable quinoxaline compound that has been shown to potently, selectively, allosterically, and reversibly inhibit Akt1, Akt2, and Akt3 activity with IC50s of 58 nM, 210 nM, and 2119 nM, respectively.
Especificaciones
Nombre del producto químico o material AKT inhibitor VIII
CAS 612847-09-3
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C34H29N7O
Cantidad 5 mg
Sinónimo AKTi-1/2
Información de solubilidad DMSO : 20 mg/mL (36.26 mM; Need ultrasonic)
SMILES O=C1NC2=CC=CC=C2N1C3CCN(CC4=CC=C(C5=NC6=CC(NC=N7)=C7C=C6N=C5C8=CC=CC=C8)C=C4)CC3
Peso molecular (g/mol) 551.64
Formula Weight (peso de la fórmula) 551.64
Porcentaje de pureza 98.93%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease