Learn More
Ajugol, MedChemExpress
Código de producto. 16489188
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16489188 10 mg 10 miligramos
16481598 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16489188 Proveedor Medchem Express N.º de proveedor HYN0914
MedChemExpress Ajugol is an iridoid glycoside that can be isolated from Sideritis germanicopolitana. Ajugol has anti-protozoal activity againt Trypanosoma b. rhodesiense with an IC50 of 31.8 μg/mL.
Ajugol is an iridoid glycoside that can be isolated from Sideritis germanicopolitana. Ajugol has anti-protozoal activity againt Trypanosoma b. rhodesiense with an IC50 of 31.8 μg/mL.
Especificaciones
Nombre del producto químico o material Ajugol
CAS 52949-83-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C15H24O9
Cantidad 10 mg
Información de solubilidad DMSO : ≥ 3.7 mg/mL (10.62 mM)
SMILES O[C@H]1[C@](O[C@H](CO)[C@@H](O)[C@@H]1O)([H])O[C@H](OC=C2)[C@@]3([H])[C@]2([H])[C@H](O)C[C@@]3(O)C
Peso molecular (g/mol) 348.35
Formula Weight (peso de la fórmula) 348.35
Porcentaje de pureza 95.0%
Grado Research
Forma física Solid