Learn More
AGN 193109, MedChemExpress
Código de producto. 16442658
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16442658 5 mg 5 miligramos
16454116 25 mg 25 miligramos
16430459 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16442658 Proveedor Medchem Express N.º de proveedor HYU00449
MedChemExpress AGN 193109 is a retinoid analog, and acts as a specific and highly effective antagonist of retinoic acid receptors (RARs), with Kds of 2 nM, 2 nM, and 3 nM for RARα, RARβ, and RARγ, respectively.
AGN 193109 is a retinoid analog, and acts as a specific and highly effective antagonist of retinoic acid receptors (RARs), with Kds of 2 nM, 2 nM, and 3 nM for RARα, RARβ, and RARγ, respectively.
Especificaciones
Nombre del producto químico o material AGN 193109
CAS 171746-21-7
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C28H24O2
Cantidad 5 mg
Información de solubilidad DMSO : 2 mg/mL (5.10 mM; Need warming)
SMILES O=C(O)C1=CC=C(C#CC2=CC=C3C(C)(C)CC=C(C4=CC=C(C)C=C4)C3=C2)C=C1
Peso molecular (g/mol) 392.49
Formula Weight (peso de la fórmula) 392.49
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death