Learn More
Clinitrol™ 290 Reference Solution, 2mL; Advanced Instruments
Critical normal human serum midpoint of 290mOsm
Marca:  Advanced Instruments 3MA029
Encompass
Código de producto. 15158616
  • 231.00€ / paquete de 10
Descripción
Critical normal human serum midpoint of 290mOsm
  • Checks performance of freezing-point osmometers in the critical area of normal serum values
  • Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
  • Pipetted into a tube and run just like normal serum
  • Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
  • Does not require mixing, reconstituting or refrigerating; one-year shelf life