missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADFP Polyclonal Antibody
GREENER_CHOICE

Código de producto. 11541832
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11541832 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11541832 Proveedor Invitrogen™ N.º de proveedor PA116972

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

The target sequence has 88% sequence homology with rat ADFP.

Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ADFP
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
génica Plin2
N.º de referencia del gen P43883, Q99541, Q9TUM6
Alias de gen AA407157; Adfp; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; Adrp; ADRP antibody; MGC10598; perilipin 2; perilipin-2; Plin2
Símbolos de los genes Plin2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide made to a C-terminal region of mouse ADFP (within residues 350-425).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11520, 123, 280981, 298199
Especies diana Human, Mouse, Rat, Bovine
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • ADFP Polyclonal Antibody
Palabra de advertencia
  • Atención
Clasificación
  • Lesión ocular grave/irritación ocular Categoría 2
Indicaciones de peligro
  • H319-Provoca irritación ocular grave.
Consejos de prudencia
  • P260-No respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
  • P264-Lavarse concienzudamente tras la manipulación.
  • P305+P351+P338-EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
Otros peligros
  • MIXTURE LIST-Contain: glycerol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.