missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAP Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-20401 can be used with blocking peptide PEP-0518.
The adhesion and degranulation adaptor protein (ADAP) was initially identified as a molecular adapter that couples T cell receptor (TCR) stimulation to the avidity of integrins governing T cell adhesion. TCR stimulation promotes the formation of a multi-protein complex containing CARMA1, MALT1, and BCL-10, which through the association of ADAP, ultimately activates the NF-kappa-B family of transcription factors. More recent experiments have shown that ADAP controls optimal T cell proliferation, cytokine production, and expression of the Bcl-2 family member Bcl-x(L), suggesting that ADAP regulates T cell activation by promoting antigen-dependent T cell-antigen presenting cell (APC) activation. At least three isoforms of ADAP are known to exist.
Specifications
Specifications
| Antígeno | ADAP |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | FYB1 |
| N.º de referencia del gen | O15117 |
| Alias de gen | ADAP; adhesion and degranulation promoting adaptor protein; adhesion and degranulation-promoting adaptor protein; B630013F22Rik; Fyb; Fyb1; FYB-120/130; FYN binding protein; FYN binding protein (FYB-120/130); FYN binding protein FYB-130; FYN-binding protein; FYN-binding protein 1; FYN-T-binding protein; p120/130; p120/p130; PRO0823; RGD1563421; Slap130; SLAP-130; SLP-76-associated phosphoprotein |
| Símbolos de los genes | FYB1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?