missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOT9 Polyclonal Antibody

Descripción
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Acyl-CoA thioesterases (ACOTs) are a group of enzymes that catalyze the hydrolysis of acyl-CoA to form coenzyme A (CoA) and a free fatty acid. Through their catalytic activity, ACOTs are able to regulate the level of fatty acids and acyl-CoAs within the cell. ACOT9 (acyl-CoA thioesterase 9), also known as ACATE2, MT-ACT48 (mitochondrial acyl-CoA thioesterase of 48 kDa) or CGI-16, is a 406 amino acid member of the acyl-CoA hydrolase protein family. ACOT9 contains a C-terminal 80-amino acid domain that is conserved from mouse to human, suggesting that the C-terminus may confer the catalytic activity of ACOT9. The gene encoding ACOT9 is located on chromosome X and the expressed ACOT9 protein is localized to the mitochondrion.
Especificaciones
Especificaciones
| Antígeno | ACOT9 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | ACOT9 |
| N.º de referencia del gen | Q9R0X4, Q9Y305 |
| Alias de gen | 0610041P13Rik; Acate2; ACOT9; Acyl coenzyme A thioester hydrolase 2; acyl-CoA thioester hydrolase 9; Acyl-CoA thioesterase 9; acyl-Coenzyme A thioesterase 2, mitochondrial; acyl-coenzyme A thioesterase 9, mitochondrial; C76421; CGI-16; Mitochondrial 48 kDa acyl-CoA thioester hydrolase 1; mitochondrial acyl-CoA thioesterase; MTACT48; MT-ACT48; mt-ACT48.1; MTE-2; p48; Protein U8; U8 |
| Símbolos de los genes | ACOT9 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?