Learn More
Acetyllovastatin, MedChemExpress
Código de producto. 16495926
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16495926 100 mg 100 miligramos
16461477 50 mg 50 miligramos
16455268 10 mg 10 miligramos
16417987 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16495926 Proveedor Medchem Express N.º de proveedor HY126237
MedChemExpress Acetyllovastatin, a acetate of Lovastatin, presentes a moderate inhibitory effect against the enzyme acetylcholinesterase with an IC50 of 79 μg/mL. Lovastatin has been found to display antifungal activity, and suppresses proliferation of a number of transformed cell lines.
Acetyllovastatin, a acetate of Lovastatin, presentes a moderate inhibitory effect against the enzyme acetylcholinesterase with an IC50 of 79 μg/mL. Lovastatin has been found to display antifungal activity, and suppresses proliferation of a number of transformed cell lines.
Especificaciones
Nombre del producto químico o material Acetyllovastatin
Forma física Solid
CAS 81189-92-6
Color Blanco
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C26H38O6
Para utilizar con (aplicación) COVID-19-immunoregulation
Información de solubilidad DMSO : ≥ 250 mg/mL (559.81 mM)
SMILES CC[C@H](C)C(O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](OC(C)=O)CC(O3)=O)[C@@]12[H])=O
Peso molecular (g/mol) 446.58
Formula Weight (peso de la fórmula) 446.58
Porcentaje de pureza 98.86%