Learn More
Conjugado de acetileno-PEG4-carboxirodamina 110, Thermo Scientific Chemicals
Código de producto. 15456629
Cambiar vista
Click to view available options
Cantidad:
5 mg
25 mg
Tamaño de la unidad:
25 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15456629 25 mg 25 miligramos
15446629 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15456629 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J64892.MB
Solicitar formato a granel o personalizado
Un reactivo de etiquetado a base de carboxirodamina 110
A carboxyrhodamine 110 based labeling reagent; reacts with azides via a copper-catalyzed click reactions. Carboxyrhodamine 110 is a bright and photostable green fluorescent label that can be excited at a wavelength of 488nm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 1615739-95-1
Fórmula molecular C32H33N3O8
Número MDL MFCD22380741
Sinónimo Acetylene-Fluor 488; Rhodamine 110-PEG4-alkyne
Especificaciones
Nombre del producto químico o material Acetylene-PEG4-carboxyrhodamine 110 conjugate
Color Rojo
Cantidad 25 mg
Información de solubilidad Soluble in DMSO,DMF,or methanol
Formula Weight (peso de la fórmula) 587.63
Forma física Sólido