missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acetyl-CoA Carboxylase Polyclonal Antibody

Código de producto. 16320305
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16320305 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16320305 Proveedor Invitrogen™ N.º de proveedor PA583343

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: MDEPSPLAQP LELNQHSRFI IGSVSEDNSE DEISNLVKLD LLEEKEGSLS PASVGSDTLS DL Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 94%.

Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Acetyl-CoA Carboxylase
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica Acaca
N.º de referencia del gen Q13085
Alias de gen A1Z784_DROME; A530025K05Rik; Acac; Acaca; ACACAD; Acc; ACC1; ACCA; ACC-alpha; ACC-PA; ACC-PB; ACC-PC; ACC-PE; ACC-PF; ACC-PG; aceto-acetyl-CoA-thiolase; acetyl CoA carboxylase; acetyl coenzyme A carboxylase; Acetyl-CoA carboxylase; Acetyl-CoA carboxylase 1; acetyl-CoA carboxylase 265; acetyl-CoA carboxylase alpha; acetyl-CoA-carboxylase; acetyl-coenzyme A carboxylase; acetyl-coenzyme A carboxylase alpha; ACoT; Biotin carboxylase; CG11198; CG11198-PA; CG11198-PB; CG11198-PC; CG11198-PE; CG11198-PF; CG11198-PG; CG8723; dACC; DmACC; Dmel\CG11198; Dmel_CG11198; EC 6.3.4.14; EC 6.4.1.2; FBgn0043811; GH12002p; Gm738; I79_006999; unnamed protein product
Símbolos de los genes Acaca
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human ACACA. Recombinant protein control fragment (Product #RP-103485).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 31
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.