Learn More
(±)-acebutolol-d7 (iso-propil-d7), CDN
Product Code. 30312201
Change view
Click to view available options
Cantidad:
0.005 g
0.01 g
Unit Size:
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30312201 0.01 g 0.01 g
30314107 0.005 g 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312201 Supplier CDN Supplier No. CDND75060.01G
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Descubra más de 4000 compuestos deuterados de alta calidad, con envío directo desde stock para mayor rapidez. Ofrecemos muchas API únicas, incluidos ingredientes farmacéuticos activos, nitrosoaminas, aminoácidos, esteroides, gases, pesticidas y ácidos grasos.
TRUSTED_SUSTAINABILITY
Specifications
Nombre del producto químico o material Acebutolol-d7
CAS 2701782-36-5
Fórmula molecular C18 D7 H21 N2 O4
Cantidad 0.01 g
Sinónimo Acebutolol-d7, (±)-Acebutolol-d7 (iso-propyl-d7), Acebutolol-D7 (N-isopropyl-D7), N-[3-Acetyl-4-[2-hydroxy-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]propoxy]phenyl]butanamide
Fórmula InChI InChI=1 S/C18H28N2O4/c1-5-6-18(23)20-14-7-8-17(16(9-14)13(4)21)24-11-15(22)10-19-12(2)3/h7-9,12,15,19,22 H,5-6,10-11H2,1-4H3,(H,20,23)/i2D3,3D3,12 D
SMILES [2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1ccc(NC(=O)CCC)cc1C(=O)C)C([2 H])([2 H])[2 H]
Nombre IUPAC N-[3-acetil-4-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ilamino)-2-hidroxipropoxi]fenil]butanamida
Peso molecular (g/mol) 343.4691
Formula Weight (peso de la fórmula) 343.2488 g/mol
Porcentaje de pureza 99 atom % D, min 98% Chemical Purity