Learn More
Tocris Bioscience™ Ac4GalNAlk
Descripción
Ac4GalNAlk is a metabolic chemical reporter (MCR) for studying glycosylation and glycoproteins. In cells expressing the engineered pyrophosphorylase mut-AGX1, Ac4GalNAlk is efficiently converted into UDP- Ac4GalNAlk which is incorporated into Ac4GalNAlk-containing glycoproteins.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-Deoxy-2-[(1-oxo-4-pentyn-1-yl)amino]-D-galactopyranose-1,3,4,6-tetraacetate |
| CAS | 1810852-60-8 |
| Cantidad | 10 mg |
| Objetivo | Glycobiology Probes |
| Fórmula molecular | C19H25NO10 |
| Pureza | 95% |