Learn More
Tocris Bioscience™ Ac-FLTD-CMK
Descripción
Ac-FLTD-CMK is a potent and selective inhibitor of caspases 1, 5 and 4 (IC50 values = 46.7 nM, 0.33 μM, 1.49 μM, respectively), and inhibits murine caspase-11 at 10 μM. Ac-FLTD-CMK inhibits cleavage of gasdermin D by caspases, suppresses pyroptosis and reduces IL-1β release in macrophages.
Especificaciones
Especificaciones
| Pureza | 95% |
| CAS | 2376255-48-8 |
| Contenido y almacenamiento | Almacene a -20 °C. |
| Cantidad | 1 mg |
| Entrada diana | Caspase Inhibitors |