Learn More
Tocris Bioscience™ ABT 639
Descripción
ABT 639 is a CaV3.2 channel blocker (IC50 = 2.3 μM). Also blocks low voltage-activated (CaV3.1) currents in native rat DRG neurons (IC50 = 7.6 μM). Exhibits little or no activity at other ion channels and receptors. Displays antinociceptive effects in a mechanical hyperalgesia model in vivo. Orally bioavailable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-[[(8aR)-hexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]carbonyl]benzenesulfonamide |
| CAS | 1235560-28-7 |
| Cantidad | 10 mg |
| Fórmula molecular | C20H20ClF2N3O3S |
| Formula Weight (peso de la fórmula) | 455.91 |
| Pureza | 98% |