Learn More
Abiraterone, MedChemExpress
Código de producto. 16490356
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
Tamaño de la unidad:
1 g
10 miligramos
100 miligramos
2 g
200 miligramos
5 g
50 miligramos
500 miligramos
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
16490356 2 g 2 g
16481946 500 mg 500 miligramos
16471336 200 mg 200 miligramos
16468168 10 mg 10 miligramos
16461959 1 g 1 g
16447926 100 mg 100 miligramos
16444077 50 mg 50 miligramos
16490618 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Código de producto. 16490356 Proveedor Medchem Express N.º de proveedor HY70013
MedChemExpress Abiraterone is a potent and irreversible CYP17A1 inhibitor with antiandrogen activity, which inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17 with IC50s of 2.5 nM and 15 nM, respectively.
Abiraterone is a potent and irreversible CYP17A1 inhibitor with antiandrogen activity, which inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17 with IC50s of 2.5 nM and 15 nM, respectively.
Especificaciones
Nombre del producto químico o material Abiraterone
CAS 154229-19-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H31NO
Cantidad 2 g
Sinónimo CB-7598
Información de solubilidad DMF : 8.75 mg/mL (25.04 mM; Need ultrasonic and warming) ∣Ethanol : 5.4 mg/mL (15.45 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (14.31 mM; Need ultrasonic)
SMILES C[C@@]12C(C3=CN=CC=C3)=CC[C@]1([C@@]4(CC=C5[C@@](C)([C@]4(CC2)[H])CC[C@@H](C5)O)[H])[H]
Peso molecular (g/mol) 349.51
Formula Weight (peso de la fórmula) 349.51
Porcentaje de pureza 95.8%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death