Learn More
Tocris Bioscience™ Abacavir hemisulfate
Reverse transcriptase inhibitor; antiretroviral
Marca:  Tocris Bioscience™ 4148/10
Encompass
Código de producto. 15796545
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
Abacavir hemisulfate is a competitive reverse transcriptase inhibitor; inhibits HIV replication. Orally active and brain penetrant. Antiretroviral agent.
Especificaciones
Especificaciones
99%
188062-50-2
10 mg
(1 S,4 R)-4-[2-Amino-6-(cyclopropylamino)-9 H-purin-9-yl]-2-cyclopentene-1-methanol hemisulfate
4°C
For Research Use Only