Learn More
Tocris Bioscience™ A 779
Descripción
A 779 is a selective Mas receptor (Ang-(1-7) receptor) antagonist. Exhibits no significant affinity for AT1 or AT2 receptors at a concentration of 1 μM. Inhibits antidiuretic effect of Ang-(1-7) in water-loaded rats. Also attenuates Monocrotaline-induced pulmonary fibrosis in rats.
Especificaciones
Especificaciones
| Pureza | 0.95 |
| CAS | 159432-28-7 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | Other Angiotensin Receptors |
| Peso molecular | Observed M.W.: 872.98 |
| Cantidad | 5 mg |
| Forma estructural | C39H60N12O11 |